KEY465196936 Key Organics / BIONET

5-(4-chlorophenyl)-2-[(4-fluorophenyl)sulfanyl]pyridine-3-carbonitrile

  • MDL number MFCD00231603
  • Cas Number : 338955-19-4
  • Molecular Weight : 340.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78717

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5J-386S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart