KEY465196954 Key Organics / BIONET

3-chloro-2-[2-(prop-2-yn-1-ylsulfanyl)-1H-imidazol-1-yl]-5-(trifluoromethyl)pyridine

  • MDL number MFCD01568357
  • Cas Number : 338956-46-0
  • Molecular Weight : 317.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78842

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5K-017_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart