KEY465196973 Key Organics / BIONET

ethyl 5-(2,6-dimethylphenoxy)-3-phenyl-1,2,4-triazine-6-carboxylate

  • MDL number MFCD00664957
  • Cas Number : 338957-41-8
  • Molecular Weight : 349.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78984

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5K-567S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart