KEY465196977 Key Organics / BIONET

ethyl 5-phenoxy-3-[3-(trifluoromethyl)phenyl]-1,2,4-triazine-6-carboxylate

  • MDL number MFCD00664963
  • Cas Number : 338957-78-1
  • Molecular Weight : 389.334

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/79036

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5K-642S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart