KEY465196988 Key Organics / BIONET

4-amino-5-(3,4-dichlorobenzoyl)-2-(phenylamino)thiophene-3-carbonitrile

  • MDL number MFCD01568665
  • Cas Number : 338959-74-3
  • Molecular Weight : 388.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/79090

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5K-941_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart