KEY465196991 Key Organics / BIONET

(E,2E)-N-(4-fluorophenyl)-2-(methoxyimino)ethanecarbohydrazonoyl cyanide

  • MDL number MFCD00663718
  • Cas Number : 338959-78-7
  • Molecular Weight : 220.207

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/79095

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5L-002_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart