KEY465197001 Key Organics / BIONET

(2E)-2-(5-amino-1,3,4-thiadiazol-2-yl)-3-(furan-2-yl)prop-2-enenitrile

  • MDL number MFCD00663728
  • Cas Number : 338959-96-9
  • Molecular Weight : 218.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/79119

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5L-030_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart