KEY465197011 Key Organics / BIONET

4-{[(1E,3E)-3-(methoxyimino)-2-nitroprop-1-en-1-yl]amino}benzonitrile

  • MDL number MFCD00664305
  • Cas Number : 338960-18-2
  • Molecular Weight : 246.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/79161

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5L-081_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart