KEY465197019 Key Organics / BIONET

6-[(4-chlorobenzenesulfonyl)methyl]-2-methyl-N-(prop-2-en-1-yl)pyrimidin-4-amine

  • MDL number MFCD00974441
  • Cas Number : 338960-56-8
  • Molecular Weight : 337.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/79204

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5L-344S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart