KEY465197020 Key Organics / BIONET

6-[(4-methylphenyl)sulfanyl]pyridine-3-carbaldehyde

  • MDL number MFCD00974894
  • Cas Number : 338960-60-4
  • Molecular Weight : 229.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/79210

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5L-350S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart