KEY465197066 Key Organics / BIONET

2-methyl-2-({[3-(trifluoromethyl)phenyl]methyl}sulfanyl)propan-1-amine

  • MDL number MFCD00975582
  • Cas Number : 338962-67-7
  • Molecular Weight : 263.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/79519

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5M-717_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart