KEY465197074 Key Organics / BIONET

(2E)-1-(2,4-dichlorophenyl)-3-(furan-2-yl)prop-2-en-1-one

  • MDL number MFCD00617575
  • Cas Number : 918151-23-2
  • Molecular Weight : 267.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/79560

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5M-939_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart