KEY465197090 Key Organics / BIONET

ethyl N-[(2E)-2-cyano-2-{[(1-hydroxy-3-phenylpropan-2-yl)amino]methylidene}acetyl]carbamate

  • MDL number MFCD01316040
  • Cas Number : 338963-01-2
  • Molecular Weight : 317.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/79602

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5N-031_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart