KEY465197098 Key Organics / BIONET

methyl 3-cyclohexyl-2-(1H-pyrrol-1-yl)propanoate

  • MDL number MFCD01316058
  • Cas Number : 338963-18-1
  • Molecular Weight : 235.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/79626

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5N-057_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart