KEY465197111 Key Organics / BIONET

N-cyclopropyl-2-{[(4-methylphenyl)methyl]sulfanyl}-5,6,7,8-tetrahydroquinazolin-4-amine

  • MDL number MFCD01443957
  • Cas Number : 338776-83-3
  • Molecular Weight : 325.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/79706

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5N-356S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart