KEY465197144 Key Organics / BIONET

3-(4-chlorophenyl)-4-(4-methoxybenzoyl)-1-methylpyrrolidine

  • MDL number MFCD01443646
  • Cas Number : 338777-13-2
  • Molecular Weight : 329.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/79824

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5N-721_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart