KEY465197152 Key Organics / BIONET

1,3-bis(4-chlorophenyl)-5-phenyl-4,5-dihydro-1H-pyrazole

  • MDL number MFCD00227402
  • Cas Number : 7245-52-5
  • Molecular Weight : 367.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/79848

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5N-755_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart