KEY465197166 Key Organics / BIONET

2-[(4-chloronaphthalen-1-yl)oxy]-N-(3-cyanothiophen-2-yl)acetamide

  • MDL number MFCD01316191
  • Cas Number : 338777-56-3
  • Molecular Weight : 342.80

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/79895

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5N-844_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart