KEY465197174 Key Organics / BIONET

1,1,1-trifluoro-3-(1,2,3,4-tetrahydroisoquinolin-2-yl)propan-2-ol

  • MDL number MFCD02083027
  • Cas Number : 478050-27-0
  • Molecular Weight : 245.245

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/79935

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5P-062_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart