KEY465197191 Key Organics / BIONET

N-(3-{hydroxy[3-(trifluoromethyl)phenyl]methyl}pyridin-2-yl)-2,2-dimethylpropanamide

  • MDL number MFCD03102827
  • Cas Number : 866038-90-6
  • Molecular Weight : 352.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/79985

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5P-346S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart