KEY465197211 Key Organics / BIONET

2-(1-phenylmethanesulfonylpyrrolidin-2-yl)-1,3-benzothiazole

  • MDL number MFCD02082924
  • Cas Number : 672925-47-2
  • Molecular Weight : 358.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80031

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5P-524S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart