KEY465197213 Key Organics / BIONET

2,6-di-tert-butyl-4-(5-sulfanyl-1,3,4-thiadiazol-2-yl)phenol

  • MDL number MFCD04125632
  • Cas Number : 130116-03-9
  • Molecular Weight : 322.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80036

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5P-531S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart