KEY465197224 Key Organics / BIONET

2-[cyano(5-formyl-1-methyl-1H-pyrrol-2-yl)methyl]benzonitrile

  • MDL number MFCD02187000
  • Cas Number : 551931-04-5
  • Molecular Weight : 249.273

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80099

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5R-0010_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart