KEY465197231 Key Organics / BIONET

1,1,1-trifluoro-3-[(1-phenyl-1H-imidazol-2-yl)sulfanyl]propan-2-yl furan-2-carboxylate

  • MDL number MFCD02661684
  • Cas Number : 691891-12-0
  • Molecular Weight : 382.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80128

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5R-0066_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart