KEY465197240 Key Organics / BIONET

2-{2-[(1Z)-2-(2,4-dinitrophenyl)cyclohexylidene]hydrazin-1-yl}-6-fluoropyridine

  • MDL number MFCD02186883
  • Cas Number : 672949-55-2
  • Molecular Weight : 373.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80154

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5R-0213_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart