KEY465197298 Key Organics / BIONET

4-[2-(3-chlorophenyl)-4-methyl-1,3-thiazol-5-yl]pyrimidin-2-amine

  • MDL number MFCD28347928
  • Cas Number : 499795-89-0
  • Molecular Weight : 302.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 177 - 179
Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/453074

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5R-0818_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart