KEY465197299 Key Organics / BIONET

4-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-5-(4-methoxyphenyl)cyclohexane-1,3-dione

  • MDL number MFCD02661775
  • Cas Number : 439110-56-2
  • Molecular Weight : 397.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80322

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5R-0822_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart