KEY465197325 Key Organics / BIONET

1-(4-chlorophenyl)-3-[4,6-dimethyl-3-(1H-pyrrol-1-yl)thieno[2,3-b]pyridin-2-yl]urea

  • MDL number MFCD04125656
  • Cas Number : 866039-18-1
  • Molecular Weight : 396.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80391

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5R-1191_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart