KEY465197331 Key Organics / BIONET

4-[4-ethyl-5-(methylsulfanyl)-4H-1,2,4-triazol-3-yl]-1-(4-methoxybenzenesulfonyl)piperidine

  • MDL number MFCD02187322
  • Cas Number : 672950-64-0
  • Molecular Weight : 396.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80416

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5R-1300_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart