KEY465197338 Key Organics / BIONET

5-(1-methylhydrazin-1-yl)-3-phenyl-6-(trifluoromethyl)-1,2,4-triazine

  • MDL number MFCD02187449
  • Cas Number : 551921-01-8
  • Molecular Weight : 269.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80451

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5R-1515_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart