KEY465197340 Key Organics / BIONET

2-{2-[2-(trifluoromethyl)-1H-1,3-benzodiazol-1-yl]acetamido}benzoic acid

  • MDL number MFCD02187156
  • Cas Number : 672950-89-9
  • Molecular Weight : 363.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80457

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/5R-1525_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart