KEY465197377 Key Organics / BIONET

5-[(E)-[1-(2-oxooxolan-3-yl)ethylidene]amino]-1H-pyrazole-4-carbonitrile

  • MDL number MFCD03012942
  • Cas Number : 1187560-14-0
  • Molecular Weight : 218.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80568

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5T-0264_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart