KEY465197394 Key Organics / BIONET

2-{2-[(1E)-[2-(2-fluorophenyl)hydrazin-1-ylidene]methyl]-1H-pyrrol-1-yl}-1,3-thiazole

  • MDL number MFCD03012744
  • Cas Number : 692287-96-0
  • Molecular Weight : 286.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80630

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5T-0610_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart