KEY465197396 Key Organics / BIONET

4-cyanophenyl 3-(4-chlorophenyl)-3-(2,2,2-trifluoroacetamido)propanoate

  • MDL number MFCD03102833
  • Cas Number : 866039-30-7
  • Molecular Weight : 396.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80639

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5T-0622_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart