KEY465197401 Key Organics / BIONET

4-[2-(4-fluorophenyl)-1,3-thiazol-5-yl]pyrimidin-2-amine

  • MDL number MFCD16170339
  • Cas Number : 1209771-56-1
  • Molecular Weight : 272.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80653

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5T-0653_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart