KEY465197406 Key Organics / BIONET

5-[(4-chlorophenyl)sulfanyl]-4-(4-methoxyphenyl)-1,3-thiazol-2-amine

  • MDL number MFCD03617515
  • Cas Number : 685108-05-8
  • Molecular Weight : 348.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80689

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5T-0849_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart