KEY465197408 Key Organics / BIONET

methyl 2-hydroxy-4-oxo-1H,4H,6H,7H,11bH-pyrido[2,1-a]isoquinoline-1-carboxylate

  • MDL number MFCD03787879
  • Cas Number : 866039-32-9
  • Molecular Weight : 273.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80693

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5T-0855_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart