KEY465197413 Key Organics / BIONET

5-(4-fluorophenyl)-3-hydroxy-4-(1H-1,2,4-triazol-1-yl)cyclohex-2-en-1-one

  • MDL number MFCD03617422
  • Cas Number : 685107-48-6
  • Molecular Weight : 273.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80710

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5T-0882_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart