KEY465197439 Key Organics / BIONET

N'-(3-methylbenzoyl)-4-pentylcyclohexane-1-carbohydrazide

  • MDL number MFCD03787757
  • Cas Number : 866039-74-9
  • Molecular Weight : 330.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80802

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5W-0246_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart