KEY465197444 Key Organics / BIONET

(2E)-3-{[2-(methoxycarbonyl)-1-benzofuran-3-yl]carbamoyl}prop-2-enoic acid

  • MDL number MFCD03787764
  • Cas Number : 866039-83-0
  • Molecular Weight : 289.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80811

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/5W-0257_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart