KEY465197447 Key Organics / BIONET

4-[4-(3-hydroxypropyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl]benzoic acid

  • MDL number MFCD03848589
  • Cas Number : 956263-78-8
  • Molecular Weight : 314.264

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80819

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5W-0267_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart