KEY465197457 Key Organics / BIONET

ethyl 4-{[3-(1H-imidazol-1-yl)propyl]amino}-1-methyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylate

  • MDL number MFCD03848594
  • Cas Number : 866040-05-3
  • Molecular Weight : 328.376

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80835

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5W-0296_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart