KEY465197461 Key Organics / BIONET

4-{[3-(1H-imidazol-1-yl)propyl]amino}-1-methyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylic acid

  • MDL number MFCD03787777
  • Cas Number : 866040-12-2
  • Molecular Weight : 300.322

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80843

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5W-0305_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart