KEY465197462 Key Organics / BIONET

3-(1H-pyrrol-1-yl)-1-benzofuran-2-carbohydrazide

  • MDL number MFCD03787779
  • Cas Number : 866040-16-6
  • Molecular Weight : 241.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(1H-pyrrol-1-yl)-1-benzofuran-2-carbohydrazide
Inchi Code 1S/C13H11N3O2/c14-15-13(17)12-11(16-7-3-4-8-16)9-5-1-2-6-10(9)18-12/h1-8H,14H2,(H,15,17)
Inchi Key NRVZIYDVWFQSJB-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80849

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/5W-0319_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart