KEY465197463 Key Organics / BIONET

1,3-diethyl 2-({[3-methyl-1-(4-methylphenyl)-1H-pyrazol-5-yl]amino}methylidene)propanedioate

  • MDL number MFCD03787780
  • Cas Number : 956574-51-9
  • Molecular Weight : 357.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80850

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/5W-0324_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart