KEY465197480 Key Organics / BIONET

methyl 1-(4-methyl-6-phenylpyrimidin-2-yl)-5-pentyl-1H-1,2,3-triazole-4-carboxylate

  • MDL number MFCD04125683
  • Cas Number : 866040-42-8
  • Molecular Weight : 365.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80902

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5W-0820_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart