KEY465197482 Key Organics / BIONET

5-[(2-chloro-6-fluorophenyl)methoxy]-2-(hydroxymethyl)-4H-pyran-4-one

  • MDL number MFCD03787790
  • Cas Number : 866040-48-4
  • Molecular Weight : 284.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80906

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5W-0828_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart