KEY465197511 Key Organics / BIONET

3-oxo-6-phenyl-2,3-dihydropyridazine-4-carboxylic acid

  • MDL number MFCD21090773
  • Cas Number : 87769-67-3
  • Molecular Weight : 216.196

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 270
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80963

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5X-0023_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart