KEY465197523 Key Organics / BIONET

ethyl 6-(ethanesulfonyl)-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylate

  • MDL number MFCD04125730
  • Cas Number : 866041-08-9
  • Molecular Weight : 299.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81011

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5X-0309_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart