KEY465197552 Key Organics / BIONET

1-(4-chlorobenzoyl)-N-methyl-4-nitro-1H-pyrazol-3-amine

  • MDL number MFCD00127539
  • Cas Number : 318517-38-3
  • Molecular Weight : 280.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81166

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/6B-020_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart